Learn More
Nickel Electro™ Lid for Stainless Steel Bucket
High quality stainless steel construction. Nickel Electro™ Lid for Stainless Steel Bucket is compatible with 8L Bucket.
Brand:  Nickel Electro™ BUCS-L080
Additional Details : Weight : 1.00000kg
Product Code. 12892024
  • 211.00 SEK / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Description
Description
  • Stainless Steel
  • Autoclavable
  • Compatible with 8L Bucket
Specifications
Specifications
Lid, Bucket
Yes
Lid
Stainless Steel
8 L Bucket
Ea.