Learn More
Nickel Electro™ Lid for 10L Stainless Steel Bucket
For use with 10L stainlees steel buckets
Brand:  Nickel Electro™ BUCS-L100
Additional Details : Weight : 1.00000kg
Product Code. 12812034
  • 326.00 SEK / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • High quality stainless steel construction
Specifications
Specifications
Lid
Yes
Lid
Bright Polished
Stainless Steel
10 L Stainless Steel Bucket
Ea.