Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand:  Ohaus™ 30517960
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 16622302
  • 756.00 SEK / Each
Estimated Shipment: 09-09-2022
to see stock.
Add to basket
Description
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
Specifications
No Certificate
100 mg
Protective Case
Stainless Steel (316)
0.005 mg