Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand: Ohaus™ 30517958
Additional Details : Weight : 0.00100kg
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
20 mg | |
Protective Case |
Stainless Steel (316) | |
0.003 mg |