Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2
Adjusted to Meet or Exceed OIML F2 Tolerances
Brand:  Ohaus™ 30390874
Additional Details : Weight : 0.00500kg
 View more versions of this product
Product Code. 16642242
  • 341.00 SEK / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
No Certificate
5 mg
Protective Case
Stainless Steel (304)
0.060 mg