Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2
Adjusted to Meet or Exceed OIML F2 Tolerances
Brand: Ohaus™ 30390867
Additional Details : Weight : 0.00100kg
Description
- Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
1 g | |
Protective Case |
Troemner Alloy 8 | |
0.300 mg |