Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2
Adjusted to Meet or Exceed OIML F2 Tolerances
253.00 SEK - 7470.00 SEK
Specifications
Certifications/Compliance No Certificate
Includes Protective Case
View More Specs
Products 23
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
16662242
View Documents Promotion Details
Ohaus™
30390876
1 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16652242
View Documents Promotion Details
Ohaus™
30390875
2 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16642242
View Documents Promotion Details
Ohaus™
30390874
5 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16632242
View Documents Promotion Details
Ohaus™
30390873
10 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16622242
View Documents Promotion Details
Ohaus™
30390872
20 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16612242
View Documents Promotion Details
Ohaus™
30390871
50 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16602242
View Documents Promotion Details
Ohaus™
30390870
100 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16692232
View Documents Promotion Details
Ohaus™
30390869
200 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16682232
View Documents Promotion Details
Ohaus™
30390868
500 mg
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16672232
View Documents Promotion Details
Ohaus™
30390867
1 g
253.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16662232
View Documents Promotion Details
Ohaus™
30390866
2 g
266.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16652232
View Documents Promotion Details
Ohaus™
30390865
5 g
276.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16642232
View Documents Promotion Details
Ohaus™
30390864
10 g
313.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16632232
View Documents Promotion Details
Ohaus™
30390863
20 g
336.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16622232
View Documents Promotion Details
Ohaus™
30390862
50 g
373.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16602232
View Documents Promotion Details
Ohaus™
30390860
200 g
600.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16612232
View Documents Promotion Details
Ohaus™
30390861
100 g
433.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16692222
View Documents Promotion Details
Ohaus™
30390859
500 g
1020.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16682222
View Documents Promotion Details
Ohaus™
30390858
1 kg
1330.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16672222
View Documents Promotion Details
Ohaus™
30390857
2 kg
1500.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16662222
View Documents Promotion Details
Ohaus™
30390856
5 kg
2745.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16652222
View Documents Promotion Details
Ohaus™
30390855
10 kg
4890.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
16642222
View Documents Promotion Details
Ohaus™
30390854
20 kg
7470.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2022
to see stock.
Add to basket
Description
Description
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
No Certificate
Protective Case