Learn More
RETSCH Stainless Steel Feed Hoppers
Brand: RETSCH 03.785.0159
7452.75 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.50000kg
Specifications
Feed Hopper | |
2.8 L | |
For V-chute | |
Stainless Steel |