Läs mer
GFL Gesellschaft Fuer Labortec™ Stainless Steel Fume Hoods for Waterbath
Vikt (metrisk):
5.7/7.0 kg
7.9/9.1 kg
Förpackningsstorlek:
Varje
Beskrivning
- Bath, removable recessed lid with holes and perforated floor above the heating element are made of stainless steel
- Adjustable water level regulator at the right-hand side of the unit is part of the standard scope of supply
- Openings in the lid are covered by sets of rings made of heat-resistant plastic material
- Temperature range from approx. 5°C above ambient to boiling point, temperature regulation through
- tension thermostat
- Four non-slip plastic feet ensure great stability
- Heating element protected by over-temperature cut-out
Specifikationer
Specifikationer
| Omgivningstemperatur | 5°C to 99.9°C |
| Controllertyp | Tension Thermostat |
| Beskrivning | Waterbath |
| För användning med (utrustning) | Fume Hoods |
| Temperaturområde | Ambient +5° to 99.9°C |
| För användning med (applikation) | Gentle steaming work with Erlenmeyer flasks, beakers, etc. |
| Material | Stainless Steel |
| Mått (D x B x H) | 300 x 670 x 155 mm |
| Bredd (metrisk) | 670 mm |
| Höjd (metrisk) | 155 mm |
| Visa mer |