Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Unions
Provide low dead volume connections for optimum chromatography
710.00 SEK - 740.00 SEK
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
15269103
|
Trajan Scientific and Medical™
103330 |
0.0625 x 0.0625 in. |
740.00 SEK
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
15209103
|
Trajan Scientific and Medical™
101270 |
0.25 x 0.25 in. |
710.00 SEK
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
- Low dead volume
- For high temperature applications
- Leak-free connections
- Does not require back ferrules
- Used with Graphite, Graphite Vespel™ or PTFE ferrules
- Suitable for glass, fused silica, metal, plastic or glass lined tubing
- Non-swaging and no tube distortion
Specifications
Stainless Steel |