Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7748-14
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10133882
  • 1985.00 SEK / Pack of 6
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Nickel-plated Brass (Hub), Stainless Steel
0.18 mm
Luer Lock Custom Needle
2, 3, or 4
Standard Tapered
28 G
0.36 mm
6/Pk.
TLL Syringes and LT or TLL Connectors