Learn More
Stainless Steel powder, -100 mesh, Type 304-L, Thermo Scientific Chemicals
468.62 SEK - 4331.16 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15460997
|
Thermo Scientific Alfa Aesar
011085.18 |
50 g |
468.62 SEK
50g |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11336098
|
Thermo Scientific Alfa Aesar
011085.36 |
500 g |
1420.05 SEK
500g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11326098
|
Thermo Scientific Alfa Aesar
011085.A3 |
2 kg |
4331.16 SEK
2kg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Stainless Steel powder, -100 mesh, Type 304-L is the most common stainless steel alloy widely used in wire weaving. It has excellent corrosion resistance and can withstand temperatures of 1100 degrees F. It has better welding characteristics for lower carbon content.