Learn More
RETSCH Stainless Steel Sieve Insert For Cyclone Mill Twister
Brand: RETSCH 03.647.0366
1909.50 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01500kg
Specifications
1Each | |
Sieve Insert | |
Cyclone Mill Twister |
0.5mm | |
Stainless Steel |