Learn More
RETSCH Stainless Steel Sieve Insert For Cyclone Mill Twister
Brand:  RETSCH 03.647.0369
11058.00 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01500kg
 View more versions of this product
Product Code. 14891621
  • 11640.00 SEK / Each
Estimated Shipment: 02-07-2024
to see stock.
Specifications
1Each
Sieve Insert
Cyclone Mill Twister
2mm