Learn More
RETSCH Stainless-Steel Test Sieve Stacks
Brand: RETSCH 60.168.000999
75786.25 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 25.00000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve Stack | |
Certificate of compliance with ISO 3310/1 or NFX 11-504, 7 Sieves (63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm) and collecting pan |
For separation, fractioning, particle size determination | |
450 mm | |
63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm | |
ISO 3310 and ASTM E 11 | |
100mm |