Learn More
RETSCH Stainless-Steel Test Sieves, 100mm Dia., in Millimeter Pore Sizes with ISO 3310/1
Brand: RETSCH 60.106.025000
2147.00 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.15000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve | |
Shaker AS 200 | |
40mm |
For separation, fractioning, particle size determination | |
100 mm | |
25mm | |
ISO 3310/1 | |
Certificate indicating Compliance |