Learn More
RETSCH Stainless Steel Intermediate Pan for Test Sieves
Collects resulting samples from test sieves. These stainless steel intermediate pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.
Brand:  RETSCH 60.220.000400
5315.25 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.90000kg
 View more versions of this product
Product Code. 11914432
  • 5595.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel
  • Compatible with multiple test sieves, based upon diameter
  • Excellent product quality due to extensive optical inspection
  • Sieve frame with a solid base
Specifications
Specifications
400 mm
Stainless Steel
65mm
Intermediate Pan
Test sieves; Vibratory Sieve Shaker AS 200 basic