Läs mer
Azpack™ Unex™ Tubing Clips
Produktkod. 11348403
Diameter (metrisk):
10 to 14 mm
13 to 17 mm
14 to 19 mm
18 to 22 mm
21 to 25 mm
26 to 30 mm
28 to 33 mm
8 to 10 mm
Förpackningsstorlek:
Paket med 5
Produktkod. 11348403
Brand: Azpack™ 11348403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Twist nut to constrict and release tubing. Azpack™ Stainless Steel Tubing Clips are designed to distribute and apply pressure evenly.
  • Stainless steel
  • Double-banded ring for even pressure
  • Nut and bolt included to apply proper compression
Specifikationer
Typ Tubing Clamp
Material Stainless Steel (H10 304, A2)
Inkluderar Tightening bolt
Diameter (metrisk) 14 to 19 mm
Typ av käke Double-banded Ring