missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12434-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
STAM (Signal transducing adaptor molecule) is an adaptor molecule involved in downstream signaling pathways mediated by growth factors and cytokines. STAM also plays an important role in endocytosis, the JAK-STAT signaling pathway, and membrane trafficking. In humans, the gene encoding STAM is located on chromosome 10.Specifications
STAM | |
Polyclonal | |
Unconjugated | |
STAM | |
STAM, STAM 1, STAM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20844, 498798, 8027 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70297, Q92783 | |
Stam | |
STAM Fusion Protein Ag3112 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |