missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAMBPL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27315-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
STAMBPL1 is zinc metalloprotease that specifically cleaves ′Lys-63′-linked polyubiquitin chains. Does not cleave ′Lys-48′-linked polyubiquitin chains.Specifications
STAMBPL1 | |
Polyclonal | |
Unconjugated | |
STAMBPL1 | |
ALMalpha, AMSH FP, AMSH like protease, AMSH LP, AMSHLP, KIAA1373, STAM binding protein like 1, STAMBPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57559 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96FJ0 | |
STAMBPL1 | |
STAMBPL1 Fusion Protein Ag26308 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |