Learn More
Fisherbrand™ Retort Stand Assembly for Cyclone Mixers
Ideal for customers who require a permanent fixing for test tubes during long term or frequent tube mixing.
Brand: Fisherbrand™
Additional Details : Weight : 0.50000kg
Description
- Ideal for customers who require a permanent fixing for test tubes during long term or frequent tube mixing
- It can also be used to reduce 'finger or hand' fatigue in orbital mixing
- Also on open tubes during mixing it offers a safer option capping the tube
- The accessory kit comprises of retort rod and clamp arm
- The retort rod conveniently screws into a fixing point on the the Cyclone Mixer case
- This accessory clamps tubes or vials onto the rubber cup for mixing
Specifications
Stand Assembly | |
Cyclone Vortex Mixer (CM-1) |