missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Phosphate, pH 6.881, NIST, Chem-Lab
For laboratory use, pH measurements, traceable to SRM from NIST
Brand: Chem Lab CL03.0601.1000
Additional Details : Weight : 1.01000kg
Description
pH = 6.984 at 0°CpH = 6.951 at 5°C
pH = 6.923 at 10°C
pH = 6.900 at 15°C
pH = 6.881 at 20°C
pH = 6.865 at 25°C
pH = 6.853 at 30°C
pH = 6.838 at 40°C
pH = 6.833 at 50°C
pH = 6.836 at 60°C
pH = 6.845 at 70°C
pH = 6.859 at 80°C
Specifications
Phosphate | |
6.881 | |
KH2PO4 + Na2HPO4 | |
NIST traceable |
1.01g/mL | |
1 L | |
PE |