missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Standard Cardboard Cryo Boxes

These Standard Cardboard Cryo Boxes store samples at ultra-low temperatures.

Brand:  Ratiolab™ 5131504

Additional Details : Weight : 5.00000kg

Product Code. 16020362

  • 450.00 SEK / Pack of 10
Estimated Shipment: 26-08-2024
to see stock.
Add to basket

Description

Description

  • Temperature-resistant down to -80°C for storing samples at ultra-low temperatures
  • Water-repellent standard coating makes the cardboard highly durable
Specifications

Specifications

Green
Cryo Box
Non-sterile
133 x 133 x 50 mm
Cardboard
10 Boxes
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers