Learn More
Macherey-Nagel™ Standard Filter Papers for Qualitative Analysis, MN 616
Manufactured from the same raw materials as ashless papers and are particularly well suited for general laboratory filtrations.
Brand:  Macherey-Nagel™ 432009
Product Code. 10044210
  • 128.00 SEK / Pack of 100
Only null left
In Stock
Description
  • Manufactured from the same raw materials as the ashless grades
  • Particularly suited for general laboratory filtrations
  • Average ash content is about 0.1%
  • The amount of α-cellulose is about 95%
Specifications
Specifications
85 g/m2
27 sec.
100/Pk.
90 mm
Disc
MN 616
0.2 mm