Läs mer
Thermo Scientific™ Standard Fluid Transfer Assembly Adaptor Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Brand:  Thermo Scientific™ SH31087.01
Produktkod. 17303920
  • Logga in för priser / Paket med 10
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Se alternativa produkter »
Beskrivning
Beskrivning
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
Approved for storage at ambient conditions (15°C to 30°C).
6.35mm (1/4 in) x 1.6mm (1/16 in) wall to 9.52mm (3/8 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2m (20 in)