Learn More
Koettermann™ Standard Individual Table, Wheels, 1500x750mm
Koettermann™ Standard Individual Table, Wheels, 1500x750mm is a mobile laboratory table with a durable worktop.
Brand: Koettermann™ 505-00027
Additional Details : Weight : 10.00000kg
Description
- Mobile table
- Topresist worktop
- Height adjustable
- One shelf
Specifications
750 mm | |
Topresist worktop | |
Mobile Table |
Adjustable | |
1 | |
1500 mm |