Learn More
Standard Precision Buffer Solution pH 7.429, Reagecon™ Product Code.: 11496404
Standard Precision Buffer Solution pH 7.429, Reagecon™
BIOPHARMA
Product Code. 11496404 Shop All Reagecon Products
500 mL, 500mL
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11496404
Brand: Reagecon 107429
Specifications
Quantity 500 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Product Code
Lot Number742924H1 Certificate TypeCertificate of Analysis Date08/07/2025 Product Code107429
Lot Number742923E1 Certificate TypeCertificate of Analysis Date04/09/2024 Product Code107429
Lot Number742922H1 Certificate TypeCertificate of Analysis Date25/07/2023 Product Code107429
Lot Number742921B1 Certificate TypeCertificate of Analysis Date27/04/2022 Product Code107429
Product Content Correction
Product Title
Standard Precision Buffer Solution pH 7.429, Reagecon™ >