missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 45 mm Shelf Height

Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE27164

Additional Details : Weight : 0.85000kg

 View more versions of this product

Product Code. 15290884

  • 990.00 SEK / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket

Description

Description

  • For use in chest freezers
  • Racks come with a locking rod to secure the cryoboxes
  • Constructed of stainless steel
Specifications

Specifications

Locking rod
Stainless Steel
135 x 92 x 376 mm
Microtiterplates (8)
Chest Frseezers
45 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers