Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 16 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27404
Additional Details : Weight : 5.30000kg
 View more versions of this product
Product Code. 15240914
  • 2610.00 SEK / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
Stainless Steel
Microtiterplates (64)
360 x 135 x 272 mm
Upright Freezers
16 x 4
16 mm