Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
1835.00 SEK - 2960.00 SEK
Specifications
Height (Metric) | 25 mm |
---|---|
For Use With (Equipment) | Upright Freezers |
Material | Stainless Steel |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
15200944
|
Tenak™
TE27470 |
Microtiterplates (72) | 15 x 5 |
2750.00 SEK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15200984
|
Tenak™
TE27584 |
Microtiterplates (70) | 10 x 7 |
2750.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15210964
|
Tenak™
TE27536 |
Microtiterplates (72) | 12 x 6 |
2820.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15210984
|
Tenak™
TE27586 |
Microtiterplates (84) | 12 x 7 |
2960.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15240934
|
Tenak™
TE27460 |
Microtiterplates (30) | 6 x 5 |
2030.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15250934
|
Tenak™
TE27464 |
Microtiterplates (50) | 10 x 5 |
2370.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15260914
|
Tenak™
TE27410 |
Microtiterplates (24) | 6 x 4 |
1835.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15260934
|
Tenak™
TE27462 |
Microtiterplates (40) | 8 x 5 |
2200.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15270914
|
Tenak™
TE27412 |
Microtiterplates (32) | 8 x 4 |
1980.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15270934
|
Tenak™
TE27466 |
Microtiterplates (60) | 12 x 5 |
2535.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15270954
|
Tenak™
TE27530 |
Microtiterplates (36) | 6 x 6 |
2185.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15280914
|
Tenak™
TE27414 |
Microtiterplates (40) | 10 x 4 |
2110.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15280954
|
Tenak™
TE27532 |
Microtiterplates (48) | 8 x 6 |
2400.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15280974
|
Tenak™
TE27580 |
Microtiterplates (42) | 6 x 7 |
2325.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
15290914
|
Tenak™
TE27416 |
Microtiterplates (44) | 11 x 4 |
2255.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15290954
|
Tenak™
TE27534 |
Microtiterplates (60) | 10 x 6 |
2605.00 SEK
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15290974
|
Tenak™
TE27582 |
Microtiterplates (56) | 8 x 7 |
2535.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
25 mm | |
Upright Freezers | |
Stainless Steel |