missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 100 mm Box Height, with Boxes

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE23220B

Additional Details : Weight : 3.20000kg

 View more versions of this product

Product Code. 15260704

  • 2025.00 SEK / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

White cardboard boxes, 9 x 9 cell dividers
Boxes (10)
Upright Freezers
100 mm (Box)
2 x 5
Stainless Steel
683 x 139 x 210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers