Learn More
Tenak™ Standard Rack for Upright Freezer, 32 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23068
Additional Details : Weight : 3.50000kg
 View more versions of this product
Product Code. 15280644
  • 1365.00 SEK / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
9 x 4
Stainless Steel
560 x 139 x 324 mm
Boxes (36)
Upright Freezers
32 mm (Box)