missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE23180

Additional Details : Weight : 1.54000kg

 View more versions of this product

Product Code. 15230684

Quantity Price
1 924.00 SEK / Each
10 836.13 SEK / Each
Estimated Shipment: 19-10-2021

Add to basket


Description

Description

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications

Specifications

2 x 4
Upright Freezers
Boxes (8)
560 x 139 x 160 mm
75 mm (Box)
Stainless Steel
Videos
SDS
Documents
Certificates