missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE23192
Additional Details : Weight : 2.75000kg
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
3 x 5 | |
Stainless Steel | |
683 x 139 x 240 mm |
Boxes (15) | |
Upright Freezers | |
75 mm (Box) |