missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE23194
Additional Details : Weight : 3.30000kg
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
4 x 5 | |
Stainless Steel | |
683 x 139 x 320 mm |
Boxes (20) | |
Upright Freezers | |
75 mm (Box) |