missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE23186

Additional Details : Weight : 3.19000kg

 View more versions of this product

Product Code. 15290684

  • 1425.00 SEK / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications

Specifications

5 x 4
Stainless Steel
560 x 139 x 400 mm
Boxes (20)
Upright Freezers
75 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers