missing translation for 'onlineSavingsMsg'
Learn More

SICCO Star-Desiccator Protect

Product Code. 10787332
Click to view available options
Volym (metrisk):
51 l
This item is not returnable. View return policy

Product Code. 10787332

Brand: SICCO V187907

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Star-Desiccator Protect orange ACRYLIC/ALUMINIUM, with: elastic gasket-lined door, 1 desiccant tray with Silica Gel, 1 mounted hygrometer, 1 magnetic lock, 4 removable shelves. Usable interior space: W 260 x H 480 x D 330 mm

  • Aluminium frame, orange panels made of acrylic glass reduce light incidence and protect against ultraviolet radiation, including four shelves made of polycarbonate, tray, hygrometer and desiccant, usable with up to 26 shelves, stackable.
  • Orange panels reduce light incidence
  • For storage of light-sensitive substances
  • Door with magnetic catch and circular rubber seal
  • Variable height shelves
  • Easy to read electronic hygrometer
  • Telescopic shelves

Specifications

Skåp stil Front-loading
Produkttyp Nonvacuum Desiccator
Volym (metrisk) 51 l
Mått (D x B x H) Inredning 260 x 480 x 330 mm
Vikt (metrisk) 7 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.