missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10487-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.Specifications
STARD5 | |
Polyclonal | |
Unconjugated | |
STARD5 | |
STARD5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170460, 80765 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9EPQ7, Q9NSY2 | |
Stard5 | |
STARD5 Fusion Protein Ag0753 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |