missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
STAT2 Antibody (STAT2/2650), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08587AF532
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifikationer
| STAT2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Recombinant full-length human STAT2 protein (Uniprot: P52630) | |
| 100 μg | |
| Cancer, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| STAT2/2650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion