Läs mer
STAT3 Rabbit, anti-Human, Unconjugated, Clone: 3H1, Proteintech
Rabbit Recombinant Monoclonal Antibody
4982.01 SEK
Specifikationer
| Antigen | STAT3 |
|---|---|
| Klona | 3H1 |
| Koncentration | 400 μg/mL |
| Användningsområden | Western Blot, ELISA |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
17291536
|
Proteintech
80149-1-RR-100UL |
100 μL |
4982.01 SEK
100µL |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Signal transducer and activator of transcription 3 (acute-phase response factor) (STAT3, synonyms: APRF, FLJ20882, MGC16063) is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. STAT3 is activated through phosphorylation in response to various cytokines and growth factors including IFNs, EGF, IL5, IL6, HGF, LIF and BMP2. STAT3 mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of STAT3.
Specifikationer
| STAT3 | |
| 400 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse | |
| Primary |
| 3H1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| 6774 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.