missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | STAT6 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896523
|
Proteintech
10257-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16886523
|
Proteintech
10257-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.Specifications
STAT6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P42226, P52633 | |
20852, 362896, 6778 | |
STAT6 Fusion Protein Ag0410 | |
Primary | |
-20°C | |
STAT6 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D12S1644, IL 4 STAT, STAT6, STAT6B, STAT6C | |
Stat6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |