Läs mer
STEMCELL Technologies Stauprimide
Beskrivning
- Stauprimide specifically inhibits the nuclear localization of NME2, which results in the suppression of c-MYC—a key regulator of pluripotency—thereby priming cells for differentiation (Zhu et al.)
- Enhances cytokine-mediated directed differentiation of mouse and human pluripotent stem cells to multiple lineages, including definitive endoderm, neural progenitors, and mesodermal derivatives such as cardiomyocytes (Zhu et al.; Tahamtani et al.)
Specifikationer
Specifikationer
| CAS | 154589-96-5 |
| Synonym | N-Benzoyl-7-oxo Staurosporine |
| Kvantitet | 1 mg |
| Molekylformel | C35H28N4O5 |
| Renhet | ≥98% |