missing translation for 'onlineSavingsMsg'
Läs mer

Stearic Acid-1-13C, TRC

Produktkod. 30419223
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419223 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30419223 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS686500100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Stearic Acid-1-13C
CAS 85541-42-0
Molekylformel 13C C17 H36 O2
Kvantitet 100 mg
Synonym Octadecanoic-1-13C acid (9CI','ACI), 1-13C-Octadecanoic acid, Stearic-1-13C acid, [1-13C]Stearic acid
InChI formel InChI=1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i18+1
LEDER CCCCCCCCCCCCCCCCC[13C](=O)O
IUPAC-namn (1^{13}C)octadecanoic acid
Molekylvikt (g/mol) 285.47
Formel vikt 285.2749
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.