Läs mer
Stearoyl-NTA, TRC
Produktkod. 30425995
Change view
Kvantitet:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425995 10 mg
30411551 1 mg
30416349 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68654010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 581064-76-8
Molekylformel C28H52N2O7
Molekylvikt (g/mol) 528.72
Synonym N2,N2-bis(Carboxymethyl)-N6-(1-oxooctadecyl)-L-lysine
IUPAC-namn (2R)-2-[bis(carboxymethyl)amino]-6-(octadecanoylamino)hexanoic acid
LEDER CCCCCCCCCCCCCCCCCC(=O)NCCCC[C@@H](N(CC(=O)O)CC(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Stearoyl-NTA
Kvantitet 10 mg
InChI formel InChI=1S/C28H52N2O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-25(31)29-21-18-17-19-24(28(36)37)30(22-26(32)33)23-27(34)35/h24H,2-23H2,1H3,(H,29,31)(H,32,33)(H,34,35)(H,36,37)/t24-/m1/s1
Formel vikt 528.3775