Learn More
Usbeck Laborgeraete™ Steel/Ceramic Wire Cloths
Product Code. 10372901
Change view
Click to view available options
Längd (metrisk):
120 mm
160 mm
Unit Size:
Varje
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Längd (metrisk) unitSize
10372901 120 mm Varje
10732702 160 mm Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10372901 Supplier Usbeck Laborgeraete™ Supplier No. 5081
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Keramiskt centrum Keramiskt centrum
Mått (L x B) 120 x 120mm
Kanter Folded
Kvantitet 1 vardera
Form Fyrkantig
Längd (metrisk) 120 mm
Material Steel, Ceramic
Typ Wire Cloth
Bredd (metrisk) 120 mm