Learn More
R&L Enterprises™ Steel Gas Cylinder Stands
Product Code. 12975961 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
146 mm dia. cylinder
178 mm dia. cylinder
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
12975961 146 mm dia. cylinder Each
12985961 178 mm dia. cylinder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12975961
Beräknad leverans: 12-01-2027
Add to basket
Add to basket
Provides sturdy, secure support for gas cylinders in the laboratory. R&L Enterprises™ Steel Gas Cylinder Stands are strongly constructed in nylon-coated steel.
  • Available in two different sizes
  • Dimension given is the maximum diameter of cylinder accomodated
Specifications
Material Steel
Holds 146 mm dia. cylinder
Product Type Gas Cylinder Stand
For Use With (Equipment) Cylinder