Learn More
RETSCH Steel Grinding Sets
Brand:  RETSCH 01.462.0002
Additional Details : Weight : 5.20000kg
 View more versions of this product
Product Code. 11994342
  • 24472.71 SEK / Each
Estimated Shipment: 04-10-2021
Add to basket
Specifications
50mL
Steel
Grinding Set
For grinding without heavy-metal contamination
For RS 200/RS 100 Steel 1,1