Learn More
Bochem™ Steel Rods
Made of zincked steel
218.00 SEK - 276.00 SEK
Specifications
Material | Steel |
---|---|
Product Type | Rod |
For Use With (Application) | For Stand Bases |
For Use With (Equipment) | Stand bases with M10 thread |
Quantity | 1 Ea. |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10089020
|
Bochem™
5153 |
1000 mm |
218.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10717171
|
Bochem™
5165 |
1500 mm |
276.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- For use with stand bases
- M10 Thread
- Zincked steel
Specifications
Steel | |
For Stand Bases | |
1 Ea. |
Rod | |
Stand bases with M10 thread |