Learn More
Usbeck Laborgeraete™ Steel Tripod Plate
401.00 SEK - 530.00 SEK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10065591
|
Usbeck Laborgeraete™
2003 |
250 mm |
530.00 SEK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10109990
|
Usbeck Laborgeraete™
2002 |
210 mm |
401.00 SEK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10310372
|
Usbeck Laborgeraete™
2000 |
180 mm |
435.00 SEK
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Specifications
Steel | |
Tripod Plate | |
1 Ea. |