Learn More
RETSCH Steel and Zirconium Oxide and Tungsten Carbide O-ring
Brand: RETSCH
2327.50 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01000kg
Specifications
O-Ring | |
1Each | |
Steel and Zirconium Oxide and Tungsten Carbide | |
For Vibratory Disc Mill RS 200 100 to 250mL Grinding Set |